Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-4-methylene-L-proline, 97%

Manufacturer:  Alfa Aesar™ H27746MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27746MD


  Check Availability
Add to cart

Specifications

Description

Applications
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

84348-38-9
227.26
(S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
C11H17NO4
MFCD01861787
N-Boc-4-methylene-L-proline

Specifications

84348-38-9
227.26
MFCD01861787
(S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
97%
C11H17NO4
110° to 114°C
250mg
N-Boc-4-methylene-L-proline

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit