Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-methylene-L-proline, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
84348-38-9
C11H17NO4
227.26
ULLGRIBXGPATMA-QMMMGPOBSA-N
n-boc-4-methylene-l-proline,s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid,1-boc-s-4-methylenepyrrolidine-2-carboxylic acid,s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester,2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid,1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s,n-boc-4-methylene-l-prolne,n-t-boc-4-methylene-l-proline,1-tert-butoxycarbonyl-4-methyleneproline
21727417
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=C)CC1C(=O)O
84348-38-9
C11H17NO4
227.26
ULLGRIBXGPATMA-QMMMGPOBSA-N
n-boc-4-methylene-l-proline,s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid,1-boc-s-4-methylenepyrrolidine-2-carboxylic acid,s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester,2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid,1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s,n-boc-4-methylene-l-prolne,n-t-boc-4-methylene-l-proline,1-tert-butoxycarbonyl-4-methyleneproline
21727417
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=C)CC1C(=O)O
Melting Point 110.0°C to 115.0°C
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble.
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-BOC-4-methylene-L-proline

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning