Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-4-oxo-L-proline, 98%

Manufacturer:  Alfa Aesar™ H27104MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27104MD


  Check Availability
Add to cart

Specifications

Description

Applications
N-Boc-4-oxo-L-proline is used in practical Synthesis of Boc-Protected cis-4-Trifluoromethyl and cis-4-Difluoromethyl-l- prolines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

84348-37-8
229.24
(S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
C10H15NO5
MFCD01860669
N-Boc-4-oxo-L-proline

Specifications

84348-37-8
229.24
MFCD01860669
(S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
98%
C10H15NO5
≈160°C
250mg
N-Boc-4-oxo-L-proline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit