Alfa Aesar™ N-Boc-4-oxo-L-proline, 98%

Manufacturer: Alfa Aesar™ H2710403

Quantity 1g

 View more versions of this product

Catalog No. AAH2710403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 84348-37-8
    Molecular Formula C10H15NO5
    Formula Weight 229.24
    MDL Number MFCD01860669
    Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
    Chemical Name or Material N-Boc-4-oxo-L-proline

    Specifications

    Formula Weight 229.24
    Molecular Formula C10H15NO5
    MDL Number MFCD01860669
    CAS 84348-37-8
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Melting Point ≈160°C
    Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
    Chemical Name or Material N-Boc-4-oxo-L-proline
    Assay 98%

    Description

    Applications
    N-Boc-4-oxo-L-proline is used in practical Synthesis of Boc-Protected cis-4-Trifluoromethyl and cis-4-Difluoromethyl-l- prolines.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.

    2° to 8°C

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit