Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-oxo-L-proline, 98%, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
84348-37-8
C10H15NO5
229.23
CKYGSXRXTIKGAJ-ZETCQYMHSA-N
n-boc-4-oxo-l-proline,s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,n-t-boc-4-oxo-l-proline,boc-4-oxo-pro-oh,n-tert-boc-4-oxo-l-proline,2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,1-tert-butoxycarbonyl-4-oxoproline,2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid,2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
11593804
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)O
84348-37-8
C10H15NO5
229.23
CKYGSXRXTIKGAJ-ZETCQYMHSA-N
n-boc-4-oxo-l-proline,s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,n-t-boc-4-oxo-l-proline,boc-4-oxo-pro-oh,n-tert-boc-4-oxo-l-proline,2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,1-tert-butoxycarbonyl-4-oxoproline,2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid,2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
11593804
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)O
Melting Point 155°C to 163°C
Quantity 250mg
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 229.23
Percent Purity 98%
Chemical Name or Material N-BOC-4-oxo-L-proline

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning