Alfa Aesar™ N-Boc-4-oxo-L-proline methyl ester, 97%

Manufacturer: Alfa Aesar™ H2778203

Quantity 1g

 View more versions of this product

Catalog No. AAH2778203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102195-80-2
    Molecular Formula C11H17NO5
    Formula Weight 243.26
    MDL Number MFCD01861778
    Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
    Chemical Name or Material N-Boc-4-oxo-L-proline methyl ester

    Specifications

    Formula Weight 243.26
    Melting Point 35° to 40°C
    MDL Number MFCD01861778
    Quantity 1g
    Recommended Storage −30° to −10°C
    Molecular Formula C11H17NO5
    CAS 102195-80-2
    Synonym (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
    Chemical Name or Material N-Boc-4-oxo-L-proline methyl ester
    Assay 97%

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit