Alfa Aesar™ N-Boc-beta-alanine, 99%

Manufacturer: Alfa Aesar™ B2252288

Quantity 2.5g

 View more versions of this product

Catalog No. AAB2252288

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3303-84-2
    Molecular Formula C8H15NO4
    Linear Formula (CH3)3CO2CNHCH2CH2CO2H
    Formula Weight 189.21
    MDL Number MFCD00037291
    Synonym Boc-beta-Ala-OH; N-tert-Butoxycarbonyl-beta-alanine
    Chemical Name or Material N-Boc-beta-alanine

    Specifications

    Formula Weight 189.21
    Molecular Formula C8H15NO4
    Linear Formula (CH3)3CO2CNHCH2CH2CO2H
    Melting Point 73° to 76°C
    Quantity 2.5g
    MDL Number MFCD00037291
    CAS 3303-84-2
    Synonym Boc-beta-Ala-OH; N-tert-Butoxycarbonyl-beta-alanine
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Boc-beta-alanine
    Assay 99%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit