Alfa Aesar™ N-Boc-cis-4-hydroxy-L-proline, 97%

Manufacturer: Alfa Aesar™ H27850MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27850MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87691-27-8
    Molecular Formula C10H17NO5
    Formula Weight 231.25
    MDL Number MFCD02094406
    Synonym Boc-cis-Hyp-OH; 1-tert-Butoxycarbonyl-(2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
    Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline

    Specifications

    Formula Weight 231.25
    Melting Point ≈146°C
    Quantity 250mg
    Molecular Formula C10H17NO5
    MDL Number MFCD02094406
    CAS 87691-27-8
    Synonym Boc-cis-Hyp-OH; 1-tert-Butoxycarbonyl-(2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
    Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit