Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-allo-isoleucine dicyclohexylammonium salt, 98%

Click to view available options
Quantity:
250mg
1g
5g
55780-92-2
C23H44N2O4
412.615
NCLUKCZVJYNXBY-JSTCDSMHSA-N
boc-d-allo-ile-oh.dcha
122173517
N-cyclohexylcyclohexanamine;(2R,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
55780-92-2
C23H44N2O4
412.615
NCLUKCZVJYNXBY-JSTCDSMHSA-N
boc-d-allo-ile-oh.dcha
122173517
N-cyclohexylcyclohexanamine;(2R,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
Formula Weight 412.62
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Boc-D-allo-isoleucine dicyclohexylammonium salt

storageNote1 : -30° to -10°C