Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-aspartic acid 4-cyclohexyl ester, 95%

Click to view available options
Quantity:
1g
5g
112898-18-7
C15H25NO6
315.366
MFCD00060741
NLPQIWFEEKQBBN-LLVKDONJSA-N
boc-d-asp ochex-oh,boc-d-aspartic acid 4-cyslohexyl ester,boc-d-asp ochx-oh,r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,2r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,boc-d-asp ochx-oh tlc,n-alpha-t-boc-d-aspartic beta-cyclohexyl ester,n-tert-butyloxycarbonyl-d-aspartic acid 4-cyslohexyl ester
7016592
(2R)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
112898-18-7
C15H25NO6
315.366
MFCD00060741
NLPQIWFEEKQBBN-LLVKDONJSA-N
boc-d-asp ochex-oh,boc-d-aspartic acid 4-cyslohexyl ester,boc-d-asp ochx-oh,r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,2r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid,boc-d-asp ochx-oh tlc,n-alpha-t-boc-d-aspartic beta-cyclohexyl ester,n-tert-butyloxycarbonyl-d-aspartic acid 4-cyslohexyl ester
7016592
(2R)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
Applications
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Quantity 1g
Formula Weight 315.36
Percent Purity 95%
Chemical Name or Material N-Boc-D-aspartic acid 4-cyclohexyl ester

TSCA : No

Recommended Storage : Ambient temperatures