Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Boc-D-aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Quantity | 5 g |
IUPAC Name | (2R)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Formula Weight | 315.36 |
Percent Purity | 95% |
Chemical Name or Material | N-Boc-D-aspartic acid 4-cyclohexyl ester |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction