Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-glutamic acid 5-cyclohexyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
133464-27-4
C16H27NO6
329.393
MFCD00076929
FDNMLANBNJDIRG-GFCCVEGCSA-N
boc-d-glu ochex-oh,r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-l-glutamic acid 5-cyclohexyl ester,2r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-d-glutamic acid gamma-cyclohexyl ester,r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoicacid,d-glutamic acid,n-1,1-dimethylethoxy carbonyl-, 5-cyclohexyl ester,d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-cyclohexyl ester
7018750
(2R)-5-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(=O)O
133464-27-4
C16H27NO6
329.393
MFCD00076929
FDNMLANBNJDIRG-GFCCVEGCSA-N
boc-d-glu ochex-oh,r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-l-glutamic acid 5-cyclohexyl ester,2r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-d-glutamic acid gamma-cyclohexyl ester,r-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoicacid,d-glutamic acid,n-1,1-dimethylethoxy carbonyl-, 5-cyclohexyl ester,d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-cyclohexyl ester
7018750
(2R)-5-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(=O)O
Formula Weight 329.39
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Boc-D-glutamic acid 5-cyclohexyl ester

storageNote1 : -30° to -10°C