Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-phenylalanine 4-nitrophenyl ester, 95%

Click to view available options
Quantity:
5g
25g
16159-70-9
C20H22N2O6
386.404
MFCD00069874
QZIWWFMMLBBICG-QGZVFWFLSA-N
boc-d-phe-onp,r-4-nitrophenyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate,boc-d-phenylalanine 4-nitrophenyl ester,4-nitrophenyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate,ambotzbaa5570,boc-d-phenylalanine-onp,d-boc-phenylalanine p-nitrophenyl ester,n-boc-d-phenylalanine 4-nitrophenyl ester,n-tert-butoxycarbonyl-d-phenylalanine 4-nitrophenyl ester,n-alpha-t-butyloxycarbonyl-d-phenylalanine p-nitrophenyl ester
7018165
(4-nitrophenyl) (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
16159-70-9
C20H22N2O6
386.404
MFCD00069874
QZIWWFMMLBBICG-QGZVFWFLSA-N
boc-d-phe-onp,r-4-nitrophenyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate,boc-d-phenylalanine 4-nitrophenyl ester,4-nitrophenyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate,ambotzbaa5570,boc-d-phenylalanine-onp,d-boc-phenylalanine p-nitrophenyl ester,n-boc-d-phenylalanine 4-nitrophenyl ester,n-tert-butoxycarbonyl-d-phenylalanine 4-nitrophenyl ester,n-alpha-t-butyloxycarbonyl-d-phenylalanine p-nitrophenyl ester
7018165
(4-nitrophenyl) (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
Formula Weight 386.4
Physical Form Solid
Percent Purity 95%
Quantity 25g
Sensitivity Moisture Sensitive
Chemical Name or Material N-Boc-D-phenylalanine 4-nitrophenyl ester

storageNote1 : -30° to -10°C