Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-D-threonine, 95%

Click to view available options
Quantity:
5g
25g
55674-67-4
C9H17NO5
219.237
MFCD00037807
LLHOYOCAAURYRL-NTSWFWBYSA-N
boc-d-thr-oh,boc-d-threonine,n-tert-butoxycarbonyl-d-threonine,n-boc-d-threonine,2r,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid,n-alpha-tert-butoxycarbonyl-d-threonine,d-threonine, n-1,1-dimethylethoxy carbonyl,pubchem14945,boc-d-thr
6992541
(2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)O
55674-67-4
C9H17NO5
219.237
MFCD00037807
LLHOYOCAAURYRL-NTSWFWBYSA-N
boc-d-thr-oh,boc-d-threonine,n-tert-butoxycarbonyl-d-threonine,n-boc-d-threonine,2r,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid,n-alpha-tert-butoxycarbonyl-d-threonine,d-threonine, n-1,1-dimethylethoxy carbonyl,pubchem14945,boc-d-thr
6992541
(2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)O
Applications
It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Beilstein 2051734
Solubility Information Slightly soluble in water.
Formula Weight 219.23
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Boc-D-threonine

TSCA : No

Recommended Storage : Ambient temperatures