Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-DL-methionine, 98%

Click to view available options
Quantity:
250mg
1g
5g
93000-03-4
C10H19NO4S
249.325
MFCD00190811
IMUSLIHRIYOHEV-UHFFFAOYSA-N
boc-dl-met-oh,boc-dl-methionine,methionine,n-1,1-dimethylethoxy carbonyl,n-tert-butoxycarbonyl methionine,2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid,2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid,n-alpha-t-boc-l-methionine,n-tert-butoxycarbonyl-d-methionine,tert-butoxycarbonyl-l-methionine,pubchem11999
102827
2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfanylbutanoic acid
CC(C)(C)OC(=O)NC(CCSC)C(=O)O
93000-03-4
C10H19NO4S
249.325
MFCD00190811
IMUSLIHRIYOHEV-UHFFFAOYSA-N
boc-dl-met-oh,boc-dl-methionine,methionine,n-1,1-dimethylethoxy carbonyl,n-tert-butoxycarbonyl methionine,2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid,2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid,n-alpha-t-boc-l-methionine,n-tert-butoxycarbonyl-d-methionine,tert-butoxycarbonyl-l-methionine,pubchem11999
102827
2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfanylbutanoic acid
CC(C)(C)OC(=O)NC(CCSC)C(=O)O
Formula Weight 249.33
Physical Form Solid
Percent Purity 98%
Quantity 250mg
Chemical Name or Material N-Boc-DL-methionine

storageNote1 : 2° to 8°C