Alfa Aesar™ N-Boc-DL-proline, 98%

Manufacturer: Alfa Aesar™ H32347MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32347MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59433-50-0
    Molecular Formula C10H17NO4
    Formula Weight 215.25
    MDL Number MFCD00520915
    Synonym (+/-)-1-Boc-pyrrolidine-2-carboxylic acid; Boc-DL-Pro-OH
    Chemical Name or Material N-Boc-DL-proline

    Specifications

    Formula Weight 215.25
    Molecular Formula C10H17NO4
    Melting Point 101° to 104°C
    MDL Number MFCD00520915
    CAS 59433-50-0
    Quantity 250mg
    Synonym (+/-)-1-Boc-pyrrolidine-2-carboxylic acid; Boc-DL-Pro-OH
    Chemical Name or Material N-Boc-DL-proline
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit