Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-DL-tert-leucine, 97%

Click to view available options
Quantity:
250mg
1g
5g
102185-35-3
C11H21NO4
231.292
MFCD00057782
LRFZIPCTFBPFLX-UHFFFAOYSA-N
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid,boc-dl-tle-oh,n-boc-t-butylglycine,2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid,2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid,2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid,boc-tbu-dl-gly-oh,boc-dl-tert-leucine,boc-dl-tbu gly-oh,n-boc-dl-tert-leucine
5046859
3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
102185-35-3
C11H21NO4
231.292
MFCD00057782
LRFZIPCTFBPFLX-UHFFFAOYSA-N
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid,boc-dl-tle-oh,n-boc-t-butylglycine,2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid,2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid,2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid,boc-tbu-dl-gly-oh,boc-dl-tert-leucine,boc-dl-tbu gly-oh,n-boc-dl-tert-leucine
5046859
3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
Formula Weight 231.29
Physical Form Solid
Percent Purity 97%
Quantity 250mg
Chemical Name or Material N-Boc-DL-tert-leucine

storageNote1 : 2° to 8°C