Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ N-Boc-gamma-aminobutyric acid, 98+%
Click to view available options
Quantity:
1g
5g
Specifications
Melting Point | 57°C to 60°C |
Quantity | 1g |
Formula Weight | 203.24 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Boc-gamma-aminobutyric acid |
Safety and Handling
EINECSNumber : 628-652-7
TSCA : No
Recommended Storage : Ambient temperatures