Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-gamma-Aminobutyric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
57294-38-9
C9H17NO4
203.24
MFCD00037313
HIDJWBGOQFTDLU-UHFFFAOYSA-N
boc-gaba-oh,n-boc-gamma-aminobutyric acid,boc-gamma-abu-oh,4-tert-butoxycarbonyl amino butanoic acid,4-boc-amino butyric acid,4-tert-butoxycarbonylaminobutyric acid,4-tert-butoxycarbonylamino butyric acid,boc-gaba,4-tert-butoxycarbonylamino butanoic acid
294894
4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCCC(=O)O
57294-38-9
C9H17NO4
203.24
MFCD00037313
HIDJWBGOQFTDLU-UHFFFAOYSA-N
boc-gaba-oh,n-boc-gamma-aminobutyric acid,boc-gamma-abu-oh,4-tert-butoxycarbonyl amino butanoic acid,4-boc-amino butyric acid,4-tert-butoxycarbonylaminobutyric acid,4-tert-butoxycarbonylamino butyric acid,boc-gaba,4-tert-butoxycarbonylamino butanoic acid
294894
4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCCC(=O)O
Melting Point 55°C
Boiling Point 78°C to 82°C (0.2mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)3COC(=O)NH(CH2)3CO2H
Formula Weight 203.24
Physical Form Powder
Percent Purity 98%
Chemical Name or Material N-BOC-γ-Aminobutyric acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.