Alfa Aesar™ N-Boc-glycine, 98+%

Manufacturer: Alfa Aesar™ A1157922

Quantity 100g

 View more versions of this product

Catalog No. AAA1157922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4530-20-5
    Molecular Formula (CH3)3COCONHCH2CO2H
    Formula Weight 175.18
    MDL Number MFCD00002690
    Beilstein 1101514
    Synonym Boc-Gly-OH; N-(tert-Butoxycarbonyl)glycine
    Chemical Name or Material N-Boc-glycine

    Specifications

    Formula Weight 175.18
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    Molecular Formula (CH3)3COCONHCH2CO2H
    Melting Point 87° to 90°C
    MDL Number MFCD00002690
    Health Hazard 1 Danger
    CAS 4530-20-5
    Beilstein 1101514
    Synonym Boc-Gly-OH; N-(tert-Butoxycarbonyl)glycine
    Chemical Name or Material N-Boc-glycine
    Assay ≥98%

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit