Alfa Aesar™ N-Boc-hexahydro-1H-azepin-4-one, 98%

Manufacturer: Alfa Aesar™ H5521403

Quantity 1g

 View more versions of this product

Catalog No. AAH5521403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 188975-88-4
    Molecular Formula C11H19NO3
    Formula Weight 213.28
    MDL Number MFCD03788435
    Synonym tert-Butyl 4-oxoazepane-1-carboxylate; tert-Butyl hexahydro-4-oxo-1H-azepine-1-carboxylate
    Chemical Name or Material N-Boc-hexahydro-1H-azepin-4-one

    Specifications

    Formula Weight 213.28
    Molecular Formula C11H19NO3
    MDL Number MFCD03788435
    CAS 188975-88-4
    Quantity 1g
    Synonym tert-Butyl 4-oxoazepane-1-carboxylate; tert-Butyl hexahydro-4-oxo-1H-azepine-1-carboxylate
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Health Hazard 1 Warning
    Chemical Name or Material N-Boc-hexahydro-1H-azepin-4-one
    Assay 98%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.

    Solubility
    Soluble in Chloroform, Dichloromethane, Ethanol and Ethyl Ether.

    Notes
    Air sensitive. Keep under 2-8°C. Store under inert gas.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit