Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-alanine 4-nitrophenyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
2483-49-0
C14H18N2O6
310.306
MFCD00038117
SUHFNHHZORGDFI-VIFPVBQESA-N
boc-ala-onp,4-nitrophenyl 2s-2-tert-butoxycarbonyl amino propanoate,l-alanine,n-1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester,boc-l-ala-onp,boc-l-alanine 4-nitrophenyl ester,n-boc-l-alanine p-nitrophenyl ester,n-boc-l-alanine 4-nitrophenyl ester,boc-ala-onp hplc,n-t-butyl-oxycarbonyl-l-alanine p-nitrophenyl ester,s-2-tert-butoxycarbonylamino-propionic acid 4-nitro-phenyl ester
7021877
(4-nitrophenyl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C(=O)OC1=CC=C(C=C1)[N+](=O)[O-])NC(=O)OC(C)(C)C
2483-49-0
C14H18N2O6
310.306
MFCD00038117
SUHFNHHZORGDFI-VIFPVBQESA-N
boc-ala-onp,4-nitrophenyl 2s-2-tert-butoxycarbonyl amino propanoate,l-alanine,n-1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester,boc-l-ala-onp,boc-l-alanine 4-nitrophenyl ester,n-boc-l-alanine p-nitrophenyl ester,n-boc-l-alanine 4-nitrophenyl ester,boc-ala-onp hplc,n-t-butyl-oxycarbonyl-l-alanine p-nitrophenyl ester,s-2-tert-butoxycarbonylamino-propionic acid 4-nitro-phenyl ester
7021877
(4-nitrophenyl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C(=O)OC1=CC=C(C=C1)[N+](=O)[O-])NC(=O)OC(C)(C)C
Beilstein 1892073
Formula Weight 310.31
Physical Form Powder
Percent Purity 95%
Color Brown
Quantity 100g
Sensitivity Light Sensitive
Chemical Name or Material N-Boc-L-alanine 4-nitrophenyl ester

storageNote1 : 2° to 8°C