Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-alaninol, 99%

Manufacturer:  Alfa Aesar™ B2239922

Quantity 100g

 View more versions of this product

Catalog No. AAB2239922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

79069-13-9
175.23
3648840
N-Boc-L-alaninol
C8H17NO3
MFCD00043121
Boc-Ala-ol; (S)-2-(Boc-amino)-1-propanol

Specifications

79069-13-9
175.23
MFCD00043121
100g
−8.6° (c=1.2 in Methanol)
99%
C8H17NO3
60° to 62°C
3648840
Boc-Ala-ol; (S)-2-(Boc-amino)-1-propanol
N-Boc-L-alaninol

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit