Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-allo-threonine dicyclohexylammonium salt, 98%

Click to view available options
Quantity:
250mg
1g
1464025-91-9
C21H40N2O5
400.56
MFCD00038976
BBZZIJOSFVOUGF-LOLRQIBTSA-N
dicyclohexylamine 2s,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoate,2s,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid; dicha
73554152
N-cyclohexylcyclohexanamine;(2S,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)O.C1CCC(CC1)NC2CCCCC2
1464025-91-9
C21H40N2O5
400.56
MFCD00038976
BBZZIJOSFVOUGF-LOLRQIBTSA-N
dicyclohexylamine 2s,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoate,2s,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid; dicha
73554152
N-cyclohexylcyclohexanamine;(2S,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)O.C1CCC(CC1)NC2CCCCC2
Formula Weight 400.56
Physical Form Solid
Percent Purity 98%
Quantity 250mg
Chemical Name or Material N-Boc-L-allo-threonine dicyclohexylammonium salt

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures