Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 1-benzyl ester, 99%

Manufacturer:  Alfa Aesar™ B2231406

Quantity 5g

 View more versions of this product

Catalog No. AAB2231406


  Check Availability
Add to cart

Specifications

Description

Applications
N-Boc-L-aspartic acid 1-benzyl ester is used as a local anesthetic and also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

30925-18-9
323.35
Boc-Asp-OBzl; N-tert-Butoxycarbonyl-L-aspartic acid 1-benzyl ester
C16H21NO6
MFCD00065563
N-Boc-L-aspartic acid 1-benzyl ester

Specifications

30925-18-9
323.35
MFCD00065563
Boc-Asp-OBzl; N-tert-Butoxycarbonyl-L-aspartic acid 1-benzyl ester
99%
C16H21NO6
95° to 97°C
5g
N-Boc-L-aspartic acid 1-benzyl ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit