Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 4-benzyl ester, 98%

Click to view available options
Quantity:
1g
5g
7536-58-5
C16H21NO6
323.345
MFCD00065564
SOHLZANWVLCPHK-LBPRGKRZSA-N
boc-asp obzl-oh,boc-l-aspartic acid 4-benzyl ester,s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,boc-l-asp obzl-oh,boc-l-aspartic acid-4-benzyl ester,4-benzyl n-boc-l-aspartate,boc-l-aspartic acid 4-benzylester,2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,boc-aspartic acid beta-benzyl ester,n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
1581888
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
7536-58-5
C16H21NO6
323.345
MFCD00065564
SOHLZANWVLCPHK-LBPRGKRZSA-N
boc-asp obzl-oh,boc-l-aspartic acid 4-benzyl ester,s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,boc-l-asp obzl-oh,boc-l-aspartic acid-4-benzyl ester,4-benzyl n-boc-l-aspartate,boc-l-aspartic acid 4-benzylester,2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,boc-aspartic acid beta-benzyl ester,n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
1581888
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
Melting Point 110°C to 112°C
Quantity 5g
Formula Weight 323.35
Percent Purity 98%
Chemical Name or Material N-Boc-L-aspartic acid 4-benzyl ester