Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-BOC-L-Aspartic acid, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Melting Point | 116.0°C to 120.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | HO2CCH2CH[NHCO2C(CH3)3]CO2H |
Quantity | 1 g |
Specific Rotation | − 6.20 |
Solubility Information | Solubility in water: slightly soluble in water. |
Formula Weight | 233.22 |
Show More |
Safety and Handling
EINECSNumber | 237-294-7 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction