Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-beta-leucine, 95%

Click to view available options
Quantity:
250mg
1g
183990-64-9
C11H21NO4
231.292
MFCD01076232
LUXMZCJCTUATDM-MRVPVSSYSA-N
boc-l-beta-homovaline,r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid,boc-beta-hoval-oh,boc-l-beta-hval-oh,r-n-boc-3-amino-4-methylpentanoicacid,r-n-boc-3-amino-4-methylpentanoic acid,3r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid,boc-beta-leu-oh,ambotzbaa6280,boc-,a-hoval-oh
2761559
(3R)-4-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)C(CC(=O)O)NC(=O)OC(C)(C)C
183990-64-9
C11H21NO4
231.292
MFCD01076232
LUXMZCJCTUATDM-MRVPVSSYSA-N
boc-l-beta-homovaline,r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid,boc-beta-hoval-oh,boc-l-beta-hval-oh,r-n-boc-3-amino-4-methylpentanoicacid,r-n-boc-3-amino-4-methylpentanoic acid,3r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid,boc-beta-leu-oh,ambotzbaa6280,boc-,a-hoval-oh
2761559
(3R)-4-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)C(CC(=O)O)NC(=O)OC(C)(C)C
Beilstein 7568192
Formula Weight 231.29
Physical Form Solid
Percent Purity 95%
Melting Point 115°C to 120°C
Quantity 1g
Chemical Name or Material N-Boc-L-beta-leucine