Alfa Aesar™ N-Boc-L-cyclohexylglycinol, 98%

Manufacturer: Alfa Aesar™ H2773206

Quantity 5g

 View more versions of this product

Catalog No. AAH2773206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107202-39-1
    Molecular Formula C13H25NO3
    Formula Weight 243.35
    MDL Number MFCD04112591
    Synonym (S)-2-(tert-Butoxycarbonylamino)-2-cyclohexylethanol
    Chemical Name or Material N-Boc-L-cyclohexylglycinol

    Specifications

    Formula Weight 243.35
    Quantity 5g
    Optical Rotation −12° (c=2 in CHCl3)
    Molecular Formula C13H25NO3
    Melting Point 83° to 87°C
    MDL Number MFCD04112591
    CAS 107202-39-1
    Synonym (S)-2-(tert-Butoxycarbonylamino)-2-cyclohexylethanol
    Chemical Name or Material N-Boc-L-cyclohexylglycinol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit