Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-glutamic acid 5-cyclohexyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
73821-97-3
C16H27NO6
329.393
MFCD00065570
FDNMLANBNJDIRG-LBPRGKRZSA-N
boc-glu ochex-oh,boc-glu ochx-oh,s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-l-glutamic acid 5-cyclohexyl ester,n-boc-l-glutamic acid 5-cyclohexyl ester,n-tert-butoxycarbonyl-l-glutamic acid 5-cyclohexyl ester,2s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-glu ochx,pubchem12179,boc-l-glutamicacid-5-cyclohexylester
7017890
(2S)-5-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(=O)O
73821-97-3
C16H27NO6
329.393
MFCD00065570
FDNMLANBNJDIRG-LBPRGKRZSA-N
boc-glu ochex-oh,boc-glu ochx-oh,s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-l-glutamic acid 5-cyclohexyl ester,n-boc-l-glutamic acid 5-cyclohexyl ester,n-tert-butoxycarbonyl-l-glutamic acid 5-cyclohexyl ester,2s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,boc-glu ochx,pubchem12179,boc-l-glutamicacid-5-cyclohexylester
7017890
(2S)-5-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(=O)O
Beilstein 3595818
Formula Weight 329.39
Physical Form Solid
Percent Purity 95%
Quantity 100g
Chemical Name or Material N-Boc-L-glutamic acid 5-cyclohexyl ester

storageNote1 : -30° to -10°C