Alfa Aesar™ N-Boc-L-glutamic acid 5-tert-butyl ester, 99%

Manufacturer: Alfa Aesar™ B2232206

Quantity 5g

 View more versions of this product

Catalog No. AAB2232206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13726-84-6
    Molecular Formula C14H25NO6
    Formula Weight 303.36
    MDL Number MFCD00038257
    Synonym Boc-Glu(OtBu)-OH; N-tert-Butoxycarbonyl-L-glutamic acid 4-tert-butyl ester
    Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester

    Specifications

    Formula Weight 303.36
    Molecular Formula C14H25NO6
    Quantity 5g
    Melting Point 110° to 112°C
    MDL Number MFCD00038257
    CAS 13726-84-6
    Synonym Boc-Glu(OtBu)-OH; N-tert-Butoxycarbonyl-L-glutamic acid 4-tert-butyl ester
    Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit