Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-isoleucine hemihydrate, 98+%

Manufacturer:  Alfa Aesar™ H3705003

Quantity 1g

 View more versions of this product

Catalog No. AAH3705003


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

204138-23-8
240.07 (231.06 Anhydrous)
N-Boc-L-isoleucine hemihydrate
C11H21NO4·0.5H2O
MFCD00672705

Specifications

204138-23-8
240.07 (231.06 Anhydrous)
MFCD00672705
N-Boc-L-isoleucine hemihydrate
C11H21NO4·0.5H2O
67° to 70°C
1g
≥98%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit