Alfa Aesar™ N-Boc-L-isoleucine hemihydrate, 98+%

Manufacturer: Alfa Aesar™ H3705006

Quantity 5g

 View more versions of this product

Catalog No. AAH3705006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 204138-23-8
    Molecular Formula C11H21NO4·0.5H2O
    Formula Weight 240.07 (231.06 Anhydrous)
    MDL Number MFCD00672705
    Chemical Name or Material N-Boc-L-isoleucine hemihydrate

    Specifications

    Formula Weight 240.07 (231.06 Anhydrous)
    TSCA TSCA
    Molecular Formula C11H21NO4·0.5H2O
    Melting Point 67° to 70°C
    MDL Number MFCD00672705
    CAS 204138-23-8
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Boc-L-isoleucine hemihydrate
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit