Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-isoleucine N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
3392-12-9
C14H22N2O6
314.34
MFCD00022585
POBDBYGSGKMZPH-UHFFFAOYNA-N
boc-val-osu,boc-l-valine hydroxysuccinimide ester,boc-l-val-osu,s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-methylbutanoate,tert-butyl s-1-2,5-dioxopyrrolidin-1-yl oxy carbonyl-2-methylpropyl carbamate,2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-valinate,bocvalosu,2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-3-methylbutanoate,t-boc-val-osu,pubchem20978
98768
(2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
CC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
3392-12-9
C14H22N2O6
314.34
MFCD00022585
POBDBYGSGKMZPH-UHFFFAOYNA-N
boc-val-osu,boc-l-valine hydroxysuccinimide ester,boc-l-val-osu,s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-methylbutanoate,tert-butyl s-1-2,5-dioxopyrrolidin-1-yl oxy carbonyl-2-methylpropyl carbamate,2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-valinate,bocvalosu,2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-3-methylbutanoate,t-boc-val-osu,pubchem20978
98768
(2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
CC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
Beilstein 1552969
Formula Weight 328.37
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 103°C to 108°C
Quantity 100g
Chemical Name or Material N-Boc-L-isoleucine N-succinimidyl ester

storageNote1 : -30° to -10°C