Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-isoleucine N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
3392-08-3
C15H24N2O6
328.37
MFCD00022585
FATJLEZSGFVHQA-CABZTGNLSA-N
boc-ile-osu,boc-l-isoleucine hydroxysuccinimide ester,boc-l-isoleucine n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate,boc-lle-osu,boc-ile-osu hplc,n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester,1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione,2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
131632401
(2,5-dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
CCC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
3392-08-3
C15H24N2O6
328.37
MFCD00022585
FATJLEZSGFVHQA-CABZTGNLSA-N
boc-ile-osu,boc-l-isoleucine hydroxysuccinimide ester,boc-l-isoleucine n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate,boc-lle-osu,boc-ile-osu hplc,n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester,1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione,2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
131632401
(2,5-dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
CCC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
Beilstein 1552969
Formula Weight 328.37
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 103°C to 108°C
Quantity 5g
Chemical Name or Material N-Boc-L-isoleucine N-succinimidyl ester

storageNote1 : -30° to -10°C