Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-L-Leucinol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
82010-31-9
C11H23NO3
217.309
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol,n-boc-l-leucinol,boc-leucinol,boc-leu-ol,tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate,n-boc l-leucinol,pubchem12195,l-leucinol, n-boc protected,n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)CC(CO)NC(=O)OC(C)(C)C
82010-31-9
C11H23NO3
217.309
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol,n-boc-l-leucinol,boc-leucinol,boc-leu-ol,tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate,n-boc l-leucinol,pubchem12195,l-leucinol, n-boc protected,n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)CC(CO)NC(=O)OC(C)(C)C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Specific Rotation -25° to -29° (20°C, 589nm) (C=2, methanol)
Quantity 1g
Color Colorless to Yellow
Assay Percent Range 97%
Formula Weight 217.31
Physical Form Viscous Liquid
Percent Purity ≥96.0%
Chemical Name or Material N-BOC-L-Leucinol
Show More Show Less