Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-phenylalanine 4-nitrophenyl ester, 97%

Click to view available options
Quantity:
25g
100g
7535-56-0
C20H22N2O6
386.404
MFCD00042811
QZIWWFMMLBBICG-KRWDZBQOSA-N
boc-phe-onp,4-nitrophenyl n-tert-butoxycarbonyl-3-phenyl-l-alaninate,4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate,l-phenylalanine,n-1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester,4-nitrophenyl 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate,ambotzbaa5870,boc-phe-4-nitrophenyl oh,boc-phe-onp hplc,n-tert butoxycarbonylphenylalanine p-nitrophenyl ester,n-tert-butoxycarbonylphenylalanine p-nitrophenyl ester
111308
(4-nitrophenyl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
7535-56-0
C20H22N2O6
386.404
MFCD00042811
QZIWWFMMLBBICG-KRWDZBQOSA-N
boc-phe-onp,4-nitrophenyl n-tert-butoxycarbonyl-3-phenyl-l-alaninate,4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate,l-phenylalanine,n-1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester,4-nitrophenyl 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate,ambotzbaa5870,boc-phe-4-nitrophenyl oh,boc-phe-onp hplc,n-tert butoxycarbonylphenylalanine p-nitrophenyl ester,n-tert-butoxycarbonylphenylalanine p-nitrophenyl ester
111308
(4-nitrophenyl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
Beilstein 1895465
Formula Weight 386.4
Physical Form Crystalline Powder
Percent Purity 97%
Color White
Melting Point 127°C to 128°C
Quantity 25g
Sensitivity Moisture Sensitive
Chemical Name or Material N-Boc-L-phenylalanine 4-nitrophenyl ester

storageNote1 : -30° to -10°C