Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-tryptophan benzyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
57229-67-1
C23H26N2O4
394.471
MFCD00190837
CEMCPAAFDOVLDW-FQEVSTJZSA-N
boc-trp-obzl,s-2-tert-butoxycarbonylamino-3-1h-indol-3-yl-propionic acid benzyl ester,boc-l-trp-obzl,n-boc-l-tryptophan benzyl ester,2-tert-butoxycarbonylamino-3-1h-indol-3-yl-propionic acid benzyl ester,nalpha-boc-tryptophan benzyl ester,boc-l-tryptophan benzylester,n-,a-boc-l-tryptophan benzyl ester,l-tryptophan,n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester,benzyl 2s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate
10318477
benzyl (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC=CC=C3
57229-67-1
C23H26N2O4
394.471
MFCD00190837
CEMCPAAFDOVLDW-FQEVSTJZSA-N
boc-trp-obzl,s-2-tert-butoxycarbonylamino-3-1h-indol-3-yl-propionic acid benzyl ester,boc-l-trp-obzl,n-boc-l-tryptophan benzyl ester,2-tert-butoxycarbonylamino-3-1h-indol-3-yl-propionic acid benzyl ester,nalpha-boc-tryptophan benzyl ester,boc-l-tryptophan benzylester,n-,a-boc-l-tryptophan benzyl ester,l-tryptophan,n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester,benzyl 2s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate
10318477
benzyl (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC=CC=C3
Formula Weight 394.47
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 140°C to 146°C
Quantity 1g
Chemical Name or Material N-Boc-L-tryptophan benzyl ester

storageNote1 : -30° to -10°C