Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-L-tryptophan methyl ester, 96%

Click to view available options
Quantity:
5g
25g
100g
33900-28-6
C17H22N2O4
318.373
MFCD01075094
QXLOVPXUZAOKBL-AWEZNQCLSA-N
boc-trp-ome,boc-l-trp-ome,boc-l-tryptophan methyl ester,s-methyl 2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate,l-tryptophan,n-1,1-dimethylethoxy carbonyl-, methyl ester,n-boc-tryptophan methyl ester,methyl 2s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate,ambotzbaa5960,boc-l-trp-ome,n-boc-l-tryptophan methyl ester
7021503
methyl (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OC
33900-28-6
C17H22N2O4
318.373
MFCD01075094
QXLOVPXUZAOKBL-AWEZNQCLSA-N
boc-trp-ome,boc-l-trp-ome,boc-l-tryptophan methyl ester,s-methyl 2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate,l-tryptophan,n-1,1-dimethylethoxy carbonyl-, methyl ester,n-boc-tryptophan methyl ester,methyl 2s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate,ambotzbaa5960,boc-l-trp-ome,n-boc-l-tryptophan methyl ester
7021503
methyl (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OC
Formula Weight 318.37
Physical Form Powder
Percent Purity 96%
Color White
Quantity 100g
Chemical Name or Material N-Boc-L-tryptophan methyl ester

storageNote1 : -30° to -10°C