Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-L-Valinol, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
79069-14-0
C10H21NO3
203.282
MFCD00082635
OOQRRYDVICNJGC-MRVPVSSYSA-N
n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
7021464
tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)C(CO)NC(=O)OC(C)(C)C
79069-14-0
C10H21NO3
203.282
MFCD00082635
OOQRRYDVICNJGC-MRVPVSSYSA-N
n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
7021464
tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)C(CO)NC(=O)OC(C)(C)C
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Density 0.9950g/mL
Boiling Point 218.0°C
Quantity 1g
Flash Point 85°C
Assay Percent Range 96%
Formula Weight 203.28
Percent Purity 96%
Chemical Name or Material N-BOC-L-Valinol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.