Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-α-BOC-N(in)-Formyl-L-tryptophan, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
47355-10-2
C17H20N2O5
332.36
IHXHBYFWSOYYTR-ZDUSSCGKSA-N
boc-trp for-oh,n-tert-butoxycarbonyl-n'-formyl-l-tryptophan,chembl63415,nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan,s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid,boc-trp cho-oh,pubchem12191,t-boc-trp cho-oh,nalpha-boc-n1-formyl-l-tryptophan
7017963
(2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN(C2=CC=CC=C21)C=O)C(=O)O
47355-10-2
C17H20N2O5
332.36
IHXHBYFWSOYYTR-ZDUSSCGKSA-N
boc-trp for-oh,n-tert-butoxycarbonyl-n'-formyl-l-tryptophan,chembl63415,nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan,s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid,boc-trp cho-oh,pubchem12191,t-boc-trp cho-oh,nalpha-boc-n1-formyl-l-tryptophan
7017963
(2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN(C2=CC=CC=C21)C=O)C(=O)O
Water 1% Max.
Specific Rotation 19.7
Melting Point 120.0°C to 123.0°C
Color White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 332.36
Physical Form Powder
Percent Purity 97%
Chemical Name or Material N-α-BOC-N(in)-Formyl-L-tryptophan
Show More Show Less