Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-N-methyl-O-benzyl-D-tyrosine, 95%

Click to view available options
Quantity:
250mg
1g
138774-98-8
C22H27NO5
385.46
MFCD02259475
FSNRGORPOYPIJC-LJQANCHMSA-N
boc-d-metyr bzl-oh,boc-n-me-d-tyr bzl-oh,d-tyrosine, n-1, 1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl,2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl methyl amino propanoic acid,ambotzbaa5560,boc-d-n-me-tyr bzl-oh,boc-n-methyl-o-benzyl-d-tyrosine,boc-nalpha-methyl-o-benzyl-d-tyrosine,n-alpha-t-butyloxycarbonyl-n-alpha-methyl-o-benzyl-d-tyrosine,d-tyrosine,n-1,1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl
7010683
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-(4-phenylmethoxyphenyl)propanoic acid
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
138774-98-8
C22H27NO5
385.46
MFCD02259475
FSNRGORPOYPIJC-LJQANCHMSA-N
boc-d-metyr bzl-oh,boc-n-me-d-tyr bzl-oh,d-tyrosine, n-1, 1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl,2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl methyl amino propanoic acid,ambotzbaa5560,boc-d-n-me-tyr bzl-oh,boc-n-methyl-o-benzyl-d-tyrosine,boc-nalpha-methyl-o-benzyl-d-tyrosine,n-alpha-t-butyloxycarbonyl-n-alpha-methyl-o-benzyl-d-tyrosine,d-tyrosine,n-1,1-dimethylethoxy carbonyl-n-methyl-o-phenylmethyl
7010683
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-(4-phenylmethoxyphenyl)propanoic acid
CC(C)(C)OC(=O)N(C)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
Formula Weight 385.46
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Boc-N-methyl-O-benzyl-D-tyrosine

storageNote1 : -30° to -10°C