Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-N-Methylethylenediamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
121492-06-6
C8H18N2O2
174.244
QYJVBVKFXDHFPQ-UHFFFAOYSA-N
n-boc-n-methylethylenediamine,n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester,n-boc-2-aminoethyl-n-methylamine,tert-butyl n-2-aminoethyl-n-methylcarbamate,n-boc-n-methyl-ethylenediamine,tert-butyl 2-aminoethyl methyl carbamate,2-n-boc-n-methylamino ethylamine,n-2-aminoethyl-n-methylcarbamicacidtert-butylester,n-boc-n-methyl ethylenediamine
2756424
tert-butyl N-(2-aminoethyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCN
121492-06-6
C8H18N2O2
174.244
QYJVBVKFXDHFPQ-UHFFFAOYSA-N
n-boc-n-methylethylenediamine,n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester,n-boc-2-aminoethyl-n-methylamine,tert-butyl n-2-aminoethyl-n-methylcarbamate,n-boc-n-methyl-ethylenediamine,tert-butyl 2-aminoethyl methyl carbamate,2-n-boc-n-methylamino ethylamine,n-2-aminoethyl-n-methylcarbamicacidtert-butylester,n-boc-n-methyl ethylenediamine
2756424
tert-butyl N-(2-aminoethyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCN
Color Colorless to Yellow
Quantity 1mL
Boiling Point 77.0°C to 79.0°C (0.4 Torr)
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point >100°C
Specific Gravity 0.975
Density 0.9750g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 174.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N-BOC-N-Methylethylenediamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger