Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-α-BOC-N-omega-Tosyl-L-arginine, 97%, contains <15% ethyl acetate, ACROS Organics™

Click to view available options
Quantity:
5g
25g
13836-37-8,141-78-6
C18H28N4O6S
428.51
WBIIPXYJAMICNU-AWEZNQCLSA-N
boc-arg tos-oh,boc-arg tos,n alpha-boc-n omega-tosyl-l-arginine,n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine,2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid,l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl,boc-arg tos oh,boc-arg ts,ambotzbaa1068,boc arg tos-oh
83767
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N
13836-37-8,141-78-6
C18H28N4O6S
428.51
WBIIPXYJAMICNU-AWEZNQCLSA-N
boc-arg tos-oh,boc-arg tos,n alpha-boc-n omega-tosyl-l-arginine,n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine,2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid,l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl,boc-arg tos oh,boc-arg ts,ambotzbaa1068,boc arg tos-oh
83767
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N
Melting Point 99°C to 100°C
Specific Rotation -3.2
Water 1% Max.
Quantity 5g
Residual Solvents 15.0% max. (EtOAc)
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 428.51
Physical Form Powder
Percent Purity 97%
Chemical Name or Material N-α-BOC-N-ω-Tosyl-L-arginine
Show More Show Less

GHS H Statement
Flammable solid.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/protective cl

GHS Signal Word: Warning

EINECSNumber : 237-549-2