Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-α-BOC-N-β-Xanthenyl-L-asparagine, 96%, Thermo Scientific™

Catalog Number p-4258791
Click to view available options
Quantity:
1 g
5 g
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
CAS Min % 95.0
CAS Max % 100.0
Melting Point 170°C to 172°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 9.5
Formula Weight 412.45
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-α-BOC-N-β-Xanthenyl-L-asparagine
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit