Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-Nortropinone, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
185099-67-6
C12H19NO3
225.29
MENILFUADYEXNU-UHFFFAOYSA-N
tert-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate,n-boc-nortropinone,n-boc-4-nortropinone,n-tert-butoxycarbonyl nortropinone,8-boc-8-azabicyclo 3.2.1 octan-3-one,3-oxo-8-aza-bicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester,t-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate,3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester,n-boc-nororopinone,8-tert-butoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one;
2794767
tert-butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
CC(C)(C)OC(=O)N1C2CCC1CC(=O)C2
185099-67-6
C12H19NO3
225.29
MENILFUADYEXNU-UHFFFAOYSA-N
tert-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate,n-boc-nortropinone,n-boc-4-nortropinone,n-tert-butoxycarbonyl nortropinone,8-boc-8-azabicyclo 3.2.1 octan-3-one,3-oxo-8-aza-bicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester,t-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate,3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester,n-boc-nororopinone,8-tert-butoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one;
2794767
tert-butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
CC(C)(C)OC(=O)N1C2CCC1CC(=O)C2
Packaging Glass Bottle
Melting Point 66.0°C to 72.0°C
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material N-Boc-Nortropinone

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning