Alfa Aesar™ N-Boc-O-benzyl-D-serine, 99%

Manufacturer: Alfa Aesar™ B2266606

Quantity 5g

 View more versions of this product

Catalog No. AAB2266606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 47173-80-8
    Molecular Formula C15H21NO5
    Formula Weight 295.31
    MDL Number MFCD00038248
    Synonym Boc-D-Ser(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-serine
    Chemical Name or Material N-Boc-O-benzyl-D-serine

    Specifications

    Formula Weight 295.31
    Molecular Formula C15H21NO5
    Melting Point 60° to 63°C
    MDL Number MFCD00038248
    Quantity 5g
    CAS 47173-80-8
    Synonym Boc-D-Ser(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-serine
    Chemical Name or Material N-Boc-O-benzyl-D-serine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit