Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-O-benzyl-D-serine, 99%

Click to view available options
Quantity:
1g
5g
47173-80-8
C15H21NO5
295.335
MFCD00038248
DMBKPDOAQVGTST-GFCCVEGCSA-N
n-boc-o-benzyl-d-serine,boc-d-ser bzl-oh,boc-o-benzyl-d-serine,o-benzyl-n-tert-butoxycarbonyl-d-serine,d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl,2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid,2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid,pubchem12444,pubchem14944,boc-d-ser obn-oh
2733693
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O
47173-80-8
C15H21NO5
295.335
MFCD00038248
DMBKPDOAQVGTST-GFCCVEGCSA-N
n-boc-o-benzyl-d-serine,boc-d-ser bzl-oh,boc-o-benzyl-d-serine,o-benzyl-n-tert-butoxycarbonyl-d-serine,d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl,2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid,2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid,pubchem12444,pubchem14944,boc-d-ser obn-oh
2733693
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O
Melting Point 60°C to 63°C
Quantity 1g
Formula Weight 295.31
Percent Purity 99%
Chemical Name or Material N-Boc-O-benzyl-D-serine