Alfa Aesar™ N-Boc-O-benzyl-L-tyrosine, 98%

Manufacturer: Alfa Aesar™ B2345503

Quantity 1g

 View more versions of this product

Catalog No. AAB2345503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2130-96-3
    Molecular Formula C21H25NO5
    Formula Weight 371.43
    MDL Number MFCD00065597
    Synonym Boc-Tyr(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-L-tyrosine
    Chemical Name or Material N-Boc-O-benzyl-L-tyrosine

    Specifications

    Formula Weight 371.43
    Molecular Formula C21H25NO5
    Melting Point 108° to 110°C
    MDL Number MFCD00065597
    CAS 2130-96-3
    Quantity 1g
    Synonym Boc-Tyr(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-L-tyrosine
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Boc-O-benzyl-L-tyrosine
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit