Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-O-ethyl-D-tyrosine, 95%

Click to view available options
Quantity:
250mg
1g
76757-92-1
C16H23NO5
309.362
MFCD00065602
NCLAAKQIHUIOIV-CYBMUJFWSA-N
boc-d-tyr et-oh,n-boc-o-ethyl-d-tyrosine,boc-o-ethyl-d-tyrosine,d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-ethyl,2r-2-tert-butoxycarbonyl amino-3-4-ethoxyphenyl propanoic acid,ambotzbaa1456,n-tert-butoxycarbonyl-o-ethyl-d-tyrosine,n-alpha-t-butyloxycarbonyl-o-ethyl-d-tyrosine,d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-ethyl,2r-2-tert-butoxycarbonylamino-3-4-ethoxyphenyl propanoic acid
7017905
(2R)-3-(4-ethoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CCOC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OC(C)(C)C
76757-92-1
C16H23NO5
309.362
MFCD00065602
NCLAAKQIHUIOIV-CYBMUJFWSA-N
boc-d-tyr et-oh,n-boc-o-ethyl-d-tyrosine,boc-o-ethyl-d-tyrosine,d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-ethyl,2r-2-tert-butoxycarbonyl amino-3-4-ethoxyphenyl propanoic acid,ambotzbaa1456,n-tert-butoxycarbonyl-o-ethyl-d-tyrosine,n-alpha-t-butyloxycarbonyl-o-ethyl-d-tyrosine,d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-ethyl,2r-2-tert-butoxycarbonylamino-3-4-ethoxyphenyl propanoic acid
7017905
(2R)-3-(4-ethoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CCOC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OC(C)(C)C
Beilstein 3622828
Formula Weight 309.36
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Boc-O-ethyl-D-tyrosine

storageNote1 : -30° to -10°C